Amidinothiourea ≥98%

Warning

Synonyms: Guanylthiourea

A15101-36 A15101-14 A15101-22 A15101-14 A15101-22 A15101-36
AAA15101-36EA 516.38 USD
AAA15101-36 AAA15101-14 AAA15101-22 CAAAA15101-22 CAAAA15101-14 CAAAA15101-36
Amidinothiourea ≥98%
Amidinothiourea
Formula: C₂H₆N₄S
Melting Pt: 160…168 °C
Storage Temperature: Ambient
MDL Number: MFCD00014472
CAS Number: 2114-02-5
EINECS: 218-308-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR