Cholesterol acetate ≥97%

Supplier: Thermo Scientific Chemicals

Synonyms: Acetic Acid Cholesterol EsterAcetyl Cholesterol

A15052-36 A15052-22 A15052-14 A15052-36 A15052-22 A15052-14
AAA15052-22EA 253.99 USD
CAAAA15052-36 CAAAA15052-14 AAA15052-22 CAAAA15052-22 AAA15052-36 AAA15052-14
Cholesterol acetate ≥97%
Cholesterol acetate
Formula: C₂₉H₄₈O₂
MW: 428.70 g/mol
Melting Pt: 112…114 °C
Storage Temperature: Ambient
MDL Number: MFCD00003636
CAS Number: 604-35-3
EINECS: 210-066-4

Order Now

Specification Test Results

Optical rotation -43° (c=5 in chloroform)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR