1-Thien-2-ylbutan-1-one 97%

Synonyms: 1-(Thiophen-2-yl)butan-1-one

A15048-18 A15048-06 A15048-18 A15048-06 A15048-09 A15048-09
AAAA15048-06EA 28.49 USD
AAAA15048-06 CAAAA15048-18 CAAAAA15048-06 CAAAA15048-09 AAA15048-09 AAA15048-18
1-Thien-2-ylbutan-1-one 97%
1-Thien-2-ylbutan-1-one
Formula: C₈H₁₀OS
MW: 154.23 g/mol
Boiling Pt: 242…244 °C
Density: 1.09
Flash Pt: 102 °C(216 °F)
Storage Temperature: Ambient
MDL Number: MFCD00022503
CAS Number: 5333-83-5
EINECS: 226-246-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR