N-Benzylideneaniline 99%

Supplier: Thermo Scientific Chemicals
Warning

A14746-22 A14746-22 A14746-14 A14746-14
AAA14746-22EA 289.99 USD
CAAAA14746-22 CAAAA14746-14 AAA14746-22 AAA14746-14
N-Benzylideneaniline 99%
N-Benzylideneaniline
Formula: C₁₃H₁₁N
MW: 181.24 g/mol
Melting Pt: 52…54 °C
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00003027
CAS Number: 538-51-2
EINECS: 208-694-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR