Crocein scarlet 3B

Supplier: Thermo Scientific Chemicals

Synonyms: Acid Red 73

A14642-30 A14642-18 A14642-30 A14642-18
AAA14642-30EA 206.99 USD
CAAAA14642-18 AAA14642-30 CAAAA14642-30 AAA14642-18
Crocein scarlet 3B
Crocein scarlet 3B

Soluble in water

Formula: C₂₂H₁₆N₄O₇S₂
MW: 556.47 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00003907
CAS Number: 5413-75-2
EINECS: 226-502-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR