3,5-Dimethyl-1H-pyrazole-1-carboxamide 98+%

Synonyms: 1-Carbamoyl-3,5-dimethylpyrazole3,5-Dimethylpyrazole-1-carboxamide

A14523-09 A14523-30 A14523-30 A14523-18 A14523-09 A14523-18
AAA14523-30EA 442.89 USD
CAAAA14523-09 AAA14523-30 CAAAA14523-18 CAAAA14523-30 AAA14523-09 AAA14523-18
3,5-Dimethyl-1H-pyrazole-1-carboxamide 98+%
3,5-Dimethyl-1H-pyrazole-1-carboxamide
Formula: C₆H₉N₃O
MW: 139.16 g/mol
Melting Pt: 112…114 °C
Storage Temperature: Ambient
MDL Number: MFCD00003126
CAS Number: 934-48-5
EINECS: 213-283-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR