3-Amino-4-bromo-5-methyl-1H-pyrazole hydrobromide 98%

Warning

A14487-14 A14487-06 A14487-03 A14487-03 A14487-06 A14487-14
AAA14487-06EA 154.8 USD
CAAAA14487-06 CAAAA14487-03 CAAAA14487-14 AAA14487-06 AAA14487-14 AAA14487-03
3-Amino-4-bromo-5-methyl-1H-pyrazole hydrobromide 98%
3-Amino-4-bromo-5-methyl-1H-pyrazole hydrobromide
Formula: C₄H₇Br₂N₃
MW: 256.94
Melting Pt: 178…181 °C
Storage Temperature: Ambient
MDL Number: MFCD00053023
CAS Number: 167683-86-5
EINECS: 000-000-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR