4-tert-Butyl-2-methylthiazole 98%
|
Warning
|
A14148-06
A14148-06
AAA14148-06EA
70.89
USD
CAAAA14148-06
AAA14148-06
4-tert-Butyl-2-methylthiazole 98%
4-tert-Butyl-2-methylthiazole
Formula:
C₈H₁₃NS MW: 155.26 Boiling Pt: 106…108 °C/70 mm Storage Temperature: Ambient |
MDL Number:
MFCD00051643 CAS Number: 15679-11-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...