5-Methylpyrazine-2-carboxylic acid 98%

Danger

A14011-03 A14011-14 A14011-06
CAAAA14011-06 CAAAA14011-03 CAAAA14011-14
5-Methylpyrazine-2-carboxylic acid 98%
5-Methylpyrazine-2-carboxylic acid
Formula: C₆H₆N₂O₂
MW: 138.13 g/mol
Melting Pt: 165…167 °C
Storage Temperature: Ambient
EINECS: 413-260-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR