Ethyl piperidin-1-ylacetate 98%

Warning

A13663-14 A13663-06 A13663-22 A13663-06 A13663-14 A13663-22
AAA13663-14EA 99.18 USD
AAA13663-14 CAAAA13663-22 AAA13663-06 AAA13663-22 CAAAA13663-14 CAAAA13663-06
Ethyl piperidin-1-ylacetate 98%
Ethyl piperidin-1-ylacetate
Formula: C₉H₁₇NO₂
Boiling Pt: 109…111 °C/25 mm
Density: 0.984
Flash Pt: 91 °C(195 °F)
Storage Temperature: Ambient
MDL Number: MFCD00134496
CAS Number: 23853-10-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR