2-(2-Chlorophenoxy)acetohydrazide 98%

Warning

A13582-06 A13582-14 A13582-06 A13582-14
AAA13582-14EA 189.81 USD
CAAAA13582-14 CAAAA13582-06 AAA13582-14 AAA13582-06
2-(2-Chlorophenoxy)acetohydrazide 98%
2-(2-Chlorophenoxy)acetohydrazide
Formula: C₈H₉ClN₂O₂
MW: 200.63 g/mol
Melting Pt: 110…113 °C
Storage Temperature: Ambient
MDL Number: MFCD00082786
CAS Number: 36304-40-2
EINECS: 000-000-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR