Ethyl 1-benzyl-4-oxopiperidine-3-carboxylate hydrochloride, hydrate 97% (max. 10% H₂O)
|
Warning
|
Synonyms:
1-Benzyl-4-oxo-3-piperidinecarboxylic Acid Ethyl Ester Hydrochloride
A13542-30
A13542-30
A13542-09
A13542-18
A13542-09
A13542-18
AAA13542-09EA
49.79
USD
CAAAA13542-09
CAAAA13542-30
AAA13542-09
AAA13542-18
AAA13542-30
CAAAA13542-18
Ethyl 1-benzyl-4-oxopiperidine-3-carboxylate hydrochloride, hydrate 97% (max. 10% H₂O)
Ethyl 1-benzyl-4-oxopiperidine-3-carboxylate hydrochloride
Formula:
C₁₅H₁₉NO₃ MW: 297.780 g/mol Melting Pt: ca 170 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00012800 CAS Number: 1454-53-1 EINECS: 215-929-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...