Tetraisopropyl methylenebisphosphonate 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Tetraisopropyl methylenebis(phosphonate)
, Tetraisopropyl methylenediphosphonate
A13484-09
A13484-18
A13484-30
AAA13484-09EA
59.49
USD
AAA13484-09
AAAA13484-18
AAA13484-30
Tetraisopropyl methylenebisphosphonate 98%
Tetraisopropyl methylenebisphosphonate
Formula:
C₁₃H₃₀O₆P₂ MW: 344.33 Boiling Pt: 122…124 °C/1.5 mm Density: 1.080 Flash Pt: 160 °C(320 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00015021 CAS Number: 1660-95-3 EINECS: 216-765-0 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...