Isonipecotamide 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Piperidine-4-carboxamide

A13442-09 A13442-30 A13442-30 A13442-18 A13442-09 A13442-18
AAAA13442-30EA 221.99 USD
CAAAAA13442-30 AAAA13442-30 CAAAA13442-18 AAA13442-09 AAA13442-18 CAAAA13442-09
Isonipecotamide 98%
Isonipecotamide
Formula: C₆H₁₂N₂O
MW: 128.18 g/mol
Melting Pt: 148…149 °C
Storage Temperature: Ambient
MDL Number: MFCD00038012
CAS Number: 39546-32-2
EINECS: 254-501-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR