Cinnamamide predominantly trans 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Cinnamamide (Predominantly trans)

A13429-22 A13429-06 A13429-22 A13429-06 A13429-14 A13429-14
AAA13429-22EA 407.99 USD
AAA13429-22 CAAAA13429-14 CAAAA13429-06 CAAAA13429-22 AAA13429-14 AAA13429-06
Cinnamamide predominantly trans 97%
Cinnamamide
Formula: C₉H₉NO
MW: 147.177 g/mol
Melting Pt: 145…149 °C
Storage Temperature: Ambient
MDL Number: MFCD00008033
CAS Number: 621-79-4
EINECS: 210-707-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR