6-Chloro-2H-chromene-3-carboxylic acid 97%
|
Warning
|
Synonyms:
6-Chloro-2H-1-benzopyran-3-carboxylic acid
A13425-03
A13425-06
A13425-03
A13425-06
AAA13425-03EA
543.5
USD
CAAAA13425-03
CAAAA13425-06
AAA13425-03
AAA13425-06
6-Chloro-2H-chromene-3-carboxylic acid 97%
6-Chloro-2H-chromene-3-carboxylic acid
Formula:
C₁₀H₇ClO₃ MW: 210.62 g/mol Melting Pt: 240…246 °C Storage Temperature: Ambient |
MDL Number:
MFCD00052362 CAS Number: 83823-06-7 EINECS: 000-000-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...