Ethyl-4-methyl-1,2,3-thiadiazole-5-carboxylate 98%
Supplier: Thermo Scientific Chemicals
Synonyms:
Ethyl 4-methyl-1,2,3-thiadiazole-5-carboxylate
A13399-06
A13399-14
A13399-03
A13399-06
A13399-03
A13399-14
AAA13399-06EA
103.99
USD
AAA13399-06
AAA13399-03
AAA13399-14
CAAAA13399-14
CAAAA13399-03
CAAAA13399-06
Ethyl-4-methyl-1,2,3-thiadiazole-5-carboxylate 98%
Ethyl-4-methyl-1,2,3-thiadiazole-5-carboxylate
Formula:
C₆H₈N₂O₂S MW: 172.21 g/mol Boiling Pt: 117…118 °C/15 mm Density: 1.265 Storage Temperature: Ambient |
MDL Number:
MFCD00052549 CAS Number: 18212-20-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...