Tri(2-thienyl)phosphine ≥98%
A13379-06
A13379-14
A13379-14
A13379-03
A13379-03
A13379-06
AAA13379-06EA
192.61
USD
AAA13379-06
AAA13379-14
AAA13379-03
CAAAA13379-14
CAAAA13379-03
CAAAA13379-06
Tri(2-thienyl)phosphine ≥98%
Tri(2-thienyl)phosphine
Formula:
C₁₂H₉PS₃ MW: 280.36 g/mol Melting Pt: 28…30 °C Storage Temperature: Ambient |
MDL Number:
MFCD00015471 CAS Number: 24171-89-9 EINECS: 246-059-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...