1-(4-Acetylphenyl)-1H-pyrrole-2,5-dione 98%

Warning

A13282-03 A13282-06 A13282-14 A13282-03 A13282-06 A13282-14
AAA13282-06EA 257.47 USD
AAA13282-06 AAA13282-03 AAA13282-14 CAAAA13282-03 CAAAA13282-14 CAAAA13282-06
1-(4-Acetylphenyl)-1H-pyrrole-2,5-dione 98%
1-(4-Acetylphenyl)-1H-pyrrole-2,5-dione
Formula: C₁₂H₉NO₃
Melting Pt: 154…158 °C
Storage Temperature: Ambient
MDL Number: MFCD00014537
CAS Number: 1082-85-5
EINECS: 214-106-1
UN: 2811
ADR: "6.1",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR