1-Cyclopentene-1-carboxylic acid 98%

Supplier: Thermo Scientific Chemicals

Warning

Synonyms: 1-Cyclopentenecarboxylic acid

A13271-03 A13271-01 A13271-06 A13271-06 A13271-03
AAAA13271-06EA 262 USD
AAAA13271-06 AAA13271-01 CAAAAA13271-06 CAAAAA13271-03 AAAA13271-03
1-Cyclopentene-1-carboxylic acid 98%
1-Cyclopentene-1-carboxylic acid
Formula: C₆H₈O₂
MW: 112.13 g/mol
Boiling Pt: 210 °C
Melting Pt: 117…121 °C
Storage Temperature: Ambient
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR