Chlorazol black E

Danger

Synonyms: Direct Black 38

A13259-18 A13259-09
AAA13259-09EA 34.11 USD
AAA13259-09 AAA13259-18
Chlorazol black E
Chlorazol black E
Formula: C₃₄H₂₇N₉O₇S₂
MW: 781.73 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00066363
CAS Number: 1937-37-7
EINECS: 217-710-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR