Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

6-Aminopenicillanic acid ≥98%

Supplier: ALFA AESAR

Purity: 98%
CAS No.: 551-16-6
Molecular Formula: C8H12N2O3S
Formula Weight: 216.26
Storage Temperature: 2°C to 8°C
Melting Point: 200° dec.
Beilstein Reference: 15080
Optical Rotation: +275° (c=1 in 0.1N HCl)
MDL No.: MFCD00005176

Formula: C8H12N2O3S
CAS Number: 551-16-6
Specification Test Results
Optical rotation +275° (c=1 in 0.1N HCl)


Log in to see your contract pricing and availability.
  Remember me on this device
 

ORDER

Expand All / Collapse All
Size Supplier No. VWR Catalog Number Unit Price Quantity
10g A13056-09 AAA13056-09 Each (10g) Retrieving Restricted

Temperature sensitive This Item is temperature sensitive and has specific temperature and storage requirements for shipping/delivery. Requires a maximum 2 day delivery.

250g A13056-30 AAA13056-30 Each (250g) Retrieving Restricted

Temperature sensitive This Item is temperature sensitive and has specific temperature and storage requirements for shipping/delivery. Requires a maximum 2 day delivery.

50g A13056-18 AAA13056-18 Each (50g) Retrieving Restricted

Temperature sensitive This Item is temperature sensitive and has specific temperature and storage requirements for shipping/delivery. Requires a maximum 2 day delivery.

restricted In order to process your orders without delay, we request that you provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:

  • State issued document with your organization's Federal Tax ID Number
  • State issued document with your organization's Resale Tax ID Number
  • City or County issued Business License
  • State Department of Health Services License
  • Any other ID issued by the State that includes the business name and address

* Please note if your account is within the State of California two of these pieces of identification will be required.
VWR will not lift restrictions for residential shipping addresses.

Expand All / Collapse All
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Melting Point 200°C dec.
Formula Weight 216.26
CAS No. 551-16-6
Purity 98%
Storage Temperature 2°C to 8°C
Molecular Formula C8H12N2O3S
MDL No. MFCD00005176
Optical Rotation +275° (c=1 in 0.1N HCl)
Merck Reference 14,458
Beilstein Reference 15080
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Select Product:

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty