N-(4-Fluorobenzyl)urea 96%

Warning

A12865-03 A12865-06 A12865-06 A12865-03
AAA12865-03EA 52.49 USD
AAA12865-03 CAAAA12865-03 AAA12865-06 CAAAA12865-06
N-(4-Fluorobenzyl)urea 96%
N-(4-Fluorobenzyl)urea
Formula: C₈H₉FN₂O
MW: 168.17 g/mol
Melting Pt: 172…175 °C
Storage Temperature: Ambient
MDL Number: MFCD00085086
CAS Number: 76523-24-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR