Deoxyanisoin 98%
Supplier: Thermo Scientific Chemicals
Synonyms:
1,2-Bis(4-methoxyphenyl)ethanone
A12816-14
A12816-22
A12816-22
A12816-14
AAA12816-22EA
123.99
USD
AAA12816-22
AAA12816-14
CAAAA12816-22
CAAAA12816-14
Deoxyanisoin 98%
Deoxyanisoin
Formula:
C₁₆H₁₆O₃ MW: 256.30 g/mol Melting Pt: 109…113 °C Storage Temperature: Ambient |
MDL Number:
MFCD00008406 CAS Number: 120-44-5 EINECS: 204-396-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...