2-Acetyl-5-cyanothiophene 97%

Supplier: Thermo Scientific Chemicals

Warning

Synonyms: 5-Acetylthiophene-2-carbonitrile

A12765-02 A12765-02
AAA12765-02EA 45.7 USD
CAAAA12765-02 AAA12765-02
2-Acetyl-5-cyanothiophene 97%
2-Acetyl-5-cyanothiophene
Formula: C₇H₅NOS
Boiling Pt: 95…97 °C/0.5 mm
Melting Pt: 76…80 °C
Storage Temperature: Ambient
MDL Number: MFCD00052314
CAS Number: 88653-55-8
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR