3-Amino-5-tert-butyl-1H-pyrazole 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 3-Amino-5-tert-butylpyrazole

A12750-06 A12750-03 A12750-14 A12750-03 A12750-14 A12750-06
AAA12750-03EA 90.89 USD
AAA12750-03 AAA12750-14 AAA12750-06 CAAAA12750-03 CAAAA12750-14 CAAAA12750-06
3-Amino-5-tert-butyl-1H-pyrazole 98%
3-Amino-5-tert-butyl-1H-pyrazole
Formula: C₇H₁₃N₃
MW: 139.20 g/mol
Melting Pt: 75…79 °C
Storage Temperature: Ambient
MDL Number: MFCD00067987
CAS Number: 82560-12-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR