4-Benzyloxy-3-methoxycinnamic acid 98%

Warning

A12558-06 A12558-03 A12558-03
AAA12558-03EA 27.22 USD
AAA12558-03 CAAAA12558-06 CAAAA12558-03
4-Benzyloxy-3-methoxycinnamic acid 98%
4-Benzyloxy-3-methoxycinnamic acid
Formula: C₁₇H₁₆O₄
MW: 284.31 g/mol
Melting Pt: 190…194 °C
Storage Temperature: Ambient
MDL Number: MFCD00052870
CAS Number: 7152-95-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR