Methyl-3-thiopheneacetate 98%

Supplier: Thermo Scientific Chemicals

Synonyms: Methyl 2-(thiophen-3-yl)acetate

A12496-06 A12496-03 A12496-06 A12496-03
AAA12496-03EA 45.69 USD
CAAAA12496-06 AAA12496-03 AAA12496-06 CAAAA12496-03
Methyl-3-thiopheneacetate 98%
Methyl-3-thiopheneacetate
Formula: C₇H₈O₂S
MW: 156.20 g/mol
Boiling Pt: 125…129 °C/25 mm
Storage Temperature: Ambient
MDL Number: MFCD00052947
CAS Number: 58414-52-1
EINECS: 261-242-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR