1-Cyanomethylpiperidine 98%

Danger

Synonyms: Piperidinoacetonitrile

A12464-06 A12464-06 A12464-14 A12464-03 A12464-14 A12464-03
AAA12464-14EA 356.31 USD
AAA12464-14 AAA12464-03 AAA12464-06 CAAAA12464-03 CAAAA12464-14 CAAAA12464-06
1-Cyanomethylpiperidine 98%
1-Cyanomethylpiperidine
Formula: C₇H₁₂N₂
MW: 124.19 g/mol
Boiling Pt: 210 °C
Melting Pt: ca 25 °C
Density: 0.96
Storage Temperature: Ambient
MDL Number: MFCD00023733
CAS Number: 3010-03-5
EINECS: 221-131-1
UN: 3276
ADR: "6.1",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR