1,3-Divinyltetramethyldisiloxane 96% cont. up to 4% 1-vinyl-3-ethyltetramethyldisiloxane
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Tetramethyl-1,3-divinyldisiloxane
, Divinyltetramethyldisiloxane
A12463-22
A12463-14
AAAA12463-14EA
74.19
USD
AAAA12463-14
AAAA12463-22
1,3-Divinyltetramethyldisiloxane 96% cont. up to 4% 1-vinyl-3-ethyltetramethyldisiloxane
1,3-Divinyltetramethyldisiloxane
Formula:
C₈H₁₈OSi₂ MW: 186.40 g/mol Boiling Pt: 139…140 °C Melting Pt: –100 °C Density: 0.811 Flash Pt: 24 °C (75 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00014933 CAS Number: 2627-95-4 EINECS: 220-099-6 UN: 1993 ADR: "3",III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...