Lead(II) tartrate 99%

Supplier: Thermo Scientific Chemicals
Danger

A12388-36 A12388-14 A12388-36 A12388-22
AAA12388-14EA 42.61 USD
AAA12388-14 AAA12388-36 AAA12388-22 CAAAA12388-36
Lead(II) tartrate 99%
Lead(II) tartrate
Formula: C₄H₄O₆Pb
MW: 355.27 g/mol
Density: 2.53
Storage Temperature: Ambient
MDL Number: MFCD00043282
CAS Number: 815-84-9
EINECS: 212-426-6
UN: 2291
ADR: "6.1",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR