1-Mesityl-2-thiourea 99%

Danger

Synonyms: N-(2,4,6-Trimethylphenyl)thiourea

A12083-14 A12083-22 A12083-06 A12083-14 A12083-06
AAA12083-06EA 40.87 USD
AAA12083-06 CAAAA12083-22 AAA12083-14 CAAAA12083-14 CAAAA12083-06
1-Mesityl-2-thiourea 99%
1-Mesityl-2-thiourea
Formula: C₁₀H₁₄N₂S
Melting Pt: 230…231 °C
Storage Temperature: Ambient
MDL Number: MFCD00051488
CAS Number: 91059-97-1
UN: 2811
ADR: "6.1",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR