2-Chloro-3-nitro-5-thiophenesulfonyl chloride 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
5-Chloro-4-nitrothiophene-2-sulphonyl chloride
A11779-06
A11779-03
AAAA11779-06EA
145.23
USD
AAAA11779-06
AAAA11779-03
2-Chloro-3-nitro-5-thiophenesulfonyl chloride 98%
2-Chloro-3-nitro-5-thiophenesulphonyl chloride
Formula:
C₄HCl₂NO₄S₂ MW: 262.09 Melting Pt: 52…54 °C Storage Temperature: Ambient |
MDL Number:
MFCD00067978 CAS Number: 58457-24-2 UN: 3261 ADR: "8",III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...