Ethyl-3-(methylthio)propionate 98%
Synonyms:
Ethyl 3-(methylthio)propanoate
A11742-36
A11742-14
A11742-14
A11742-22
A11742-22
A11742-36
AAA11742-22EA
90.82
USD
CAAAA11742-36
CAAAA11742-14
AAA11742-22
AAA11742-14
AAA11742-36
CAAAA11742-22
Ethyl-3-(methylthio)propionate 98%
Ethyl 3-(methylthio)propionate
Formula:
C₆H₁₂O₂S MW: 148.23 g/mol Boiling Pt: 197 °C Density: 1.030 Flash Pt: 79 °C(174 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00039900 CAS Number: 13327-56-5 EINECS: 236-370-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...