2-Propylthiazole 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A11574-14
A11574-06
A11574-06
A11574-14
AAA11574-06EA
105.99
USD
AAA11574-06
CAAAA11574-14
CAAAA11574-06
AAA11574-14
2-Propylthiazole 99%
2-Propylthiazole
Formula:
C₆H₉NS MW: 127.20 Boiling Pt: 62 °C/15 mm Density: 1.03 Flash Pt: 54 °C(129 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00053154 CAS Number: 17626-75-4 EINECS: 241-606-7 UN: 1993 ADR: "3",III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...