Guanine hydrochloride 99%
Synonyms:
2-Amino-1H-purin-6(7H)-one hydrochloride
A11532-18
A11532-18
A11532-30
A11532-09
A11532-09
AAA11532-18EA
194.88
USD
CAAAAA11532-30
AAA11532-18
AAA11532-09
CAAAA11532-09
CAAAA11532-18
Guanine hydrochloride 99%
Guanine hydrochloride
Formula:
C₅H₆ClN₅O MW: 187.59 g/mol Melting Pt: >300 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00213670 CAS Number: 635-39-2 EINECS: 211-235-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...