Methyldiphenylphosphine oxide 98+%

Warning

A11484-14 A11484-06 A11484-06 A11484-14
AAAA11484-06EA 62.03 USD
AAAA11484-06 AAAA11484-14 CAAAAA11484-06 CAAAAA11484-14
Methyldiphenylphosphine oxide 98+%
Methyldiphenylphosphine oxide
Formula: C₁₃H₁₃OP
MW: 216.22 g/mol
Melting Pt: 111…114 °C
Storage Temperature: Ambient
MDL Number: MFCD00002081
CAS Number: 2129-89-7
EINECS: 218-346-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR