3-Amino-2-phenylindenone 98%

A11397-06 A11397-06 A11397-14 A11397-14
AAA11397-06EA 71.93 USD
AAA11397-06 AAA11397-14 CAAAA11397-14 CAAAA11397-06
3-Amino-2-phenylindenone 98%
3-Amino-2-phenylindenone
Formula: C₁₅H₁₁NO
Melting Pt: 268…274 °C
Storage Temperature: Ambient
MDL Number: MFCD00051614
CAS Number: 1947-47-3
EINECS: 000-000-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR