N,N-Dimethylformamide dibenzyl acetal tech. 80%, Technical Grade

Warning

A11281-09 A11281-30 A11281-18 A11281-18 A11281-09 A11281-30
AAA11281-30EA 1769.24 USD
AAA11281-30 CAAAA11281-30 CAAAA11281-09 AAA11281-18 CAAAA11281-18 AAA11281-09
N,N-Dimethylformamide dibenzyl acetal tech. 80%, Technical Grade
N,N-Dimethylformamide dibenzyl acetal
Formula: C₁₇H₂₁NO₂
Boiling Pt: 138…140 °C/0.5 mm
Density: 1,045
Storage Temperature: Ambient
MDL Number: MFCD00014869
CAS Number: 2016-04-8
EINECS: 217-946-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR