Methyl acetopyruvate 98%
Synonyms:
Methyl 2,4-dioxopentanoate
A11190-06
A11190-14
A11190-14
A11190-06
AAA11190-14EA
213.8
USD
AAA11190-14
CAAAA11190-06
CAAAA11190-14
AAA11190-06
Methyl acetopyruvate 98%
Methyl acetopyruvate
Formula:
C₆H₈O₄ MW: 144.13 g/mol Melting Pt: 60…63 °C Storage Temperature: Ambient |
MDL Number:
MFCD00043638 CAS Number: 20577-61-1 EINECS: 243-891-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...