2-Amino-5-chlorobenzamide 98+%

Supplier: Thermo Scientific Chemicals
Warning

A11153-06 A11153-06 A11153-03 A11153-03
AAA11153-03EA 50.29 USD
AAA11153-03 AAA11153-06 CAAAA11153-03 CAAAA11153-06
2-Amino-5-chlorobenzamide 98+%
2-Amino-5-chlorobenzamide
Formula: C₇H₇ClN₂O
MW: 170.60 g/mol
Melting Pt: 169…173 °C
Storage Temperature: Ambient
MDL Number: MFCD00017126
CAS Number: 5202-85-7
EINECS: 225-990-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR