3-(2-Thienyl)-1-(p-tolyl)-2-propen-1-one 98%

A11083-14 A11083-06 A11083-14 A11083-06
AAA11083-14EA 172.05 USD
AAA11083-14 CAAAA11083-14 CAAAA11083-06 AAA11083-06
3-(2-Thienyl)-1-(p-tolyl)-2-propen-1-one 98%
3-(2-Thienyl)-1-(p-tolyl)-2-propen-1-one
Formula: C₁₄H₁₂OS
MW: 228.32 g/mol
Melting Pt: 78…80 °C
Storage Temperature: Ambient
MDL Number: MFCD00022532
CAS Number: 6028-89-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR