2-Hydroxyethyl phenyl sulfide 99%

Warning

Synonyms: 2-(Phenylthio)ethanol

A10861-09 A10861-18 A10861-09 A10861-18
AAA10861-09EA 34.38 USD
CAAAA10861-18 CAAAA10861-09 AAA10861-09 AAA10861-18
2-Hydroxyethyl phenyl sulfide 99%
2-Hydroxyethyl phenyl sulphide
Formula: C₈H₁₀OS
MW: 154.23 g/mol
Boiling Pt: 115…116 °C/2 mm
Density: 1.147
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00002905
CAS Number: 699-12-7
EINECS: 211-828-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR