3,5-Diiodo-L-tyrosine dihydrate, (max. 10% H₂O) 98% (dry weight)

Supplier: Thermo Scientific Chemicals
Warning

A10702-14 A10702-06 A10702-06 A10702-14 A10702-22 A10702-22
AAA10702-14EA 99.79 USD
CAAAA10702-14 CAAAA10702-06 AAA10702-14 AAA10702-22 AAA10702-06 CAAAA10702-22
3,5-Diiodo-L-tyrosine dihydrate, (max. 10% H₂O) 98% (dry weight)
3,5-Diiodo-L-tyrosine dihydrate
Formula: C₉H₉I₂NO₃
MW: 432.98 g/mol
Melting Pt: 185 °C (dec.)
Storage Temperature: Refrigerator
MDL Number: MFCD00150275
CAS Number: 18835-59-1
EINECS: 206-092-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR