7-Aminocephalosporanic acid, (max. 2% H₂O) 98% (dry weight)
|
Danger
|
Synonyms:
7-ACA
A10530-03
A10530-03
A10530-06
A10530-06
AAA10530-06EA
126.16
USD
AAA10530-06
CAAAA10530-06
CAAAA10530-03
AAA10530-03
7-Aminocephalosporanic acid, (max. 2% H₂O) 98% (dry weight)
7-Aminocephalosporanic acid
Formula:
C₁₀H₁₂N₂O₅S MW: 272.28 g/mol Melting Pt: >300 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00005177 CAS Number: 957-68-6 EINECS: 213-485-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...