4-Nitrocinnamyl alcohol 98%

Warning

A10338-14 A10338-03 A10338-06 A10338-14 A10338-03 A10338-06
AAA10338-06EA 267.39 USD
AAA10338-06 AAA10338-14 AAA10338-03 CAAAA10338-14 CAAAA10338-03 CAAAA10338-06
4-Nitrocinnamyl alcohol 98%
4-Nitrocinnamyl alcohol
Formula: C₉H₉NO₃
Melting Pt: 126…130 °C
Storage Temperature: Ambient
MDL Number: MFCD00017045
CAS Number: 1504-63-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR