3-Chlorobenzylideneacetone 98%

Supplier: Thermo Scientific Chemicals

Synonyms: 4-(3-Chlorophenyl)but-3-en-2-one

A10332-06 A10332-14 A10332-06 A10332-14
AAA10332-14EA 333 USD
AAA10332-14 AAA10332-06 CAAAA10332-06 CAAAA10332-14
3-Chlorobenzylideneacetone 98%
3-Chlorobenzylideneacetone
Formula: C₁₀H₉ClO
MW: 180.64
Boiling Pt: 123…125 °C/1 mm
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00052851
CAS Number: 20766-36-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR