2,4-Diamino-6-hydroxy-5-nitrosopyrimidine 97%
|
|
A10147-36
A10147-22
A10147-14
A10147-22
A10147-14
A10147-36
AAA10147-14EA
46.54
USD
AAA10147-14
AAA10147-36
AAA10147-22
CAAAA10147-36
CAAAA10147-14
CAAAA10147-22
2,4-Diamino-6-hydroxy-5-nitrosopyrimidine 97%
2,4-Diamino-6-hydroxy-5-nitrosopyrimidine
Formula:
C₄H₅N₅O₂ MW: 155.12 g/mol Melting Pt: >360 °C Storage Temperature: Ambient |
MDL Number:
MFCD00023263 CAS Number: 2387-48-6 EINECS: 219-214-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...