Diphenhydramine hydrochloride 99%

Warning

Synonyms: 2-Diphenylmethoxy-N,N-dimethylethylamine Hydrochloride2-(Benzhydryloxy)-N,N-dimethylethylamine Hydrochloride

A10136-0B A10136-0B A10136-18 A10136-30 A10136-30 A10136-18
AAA10136-18EA 44.02 USD
AAA10136-18 CAAAA10136-18 AAA10136-30 CAAAA10136-0B AAA10136-0B CAAAA10136-30
Diphenhydramine hydrochloride 99%
Diphenhydramine hydrochloride

A histamine H1 receptor antagonist

Formula: C₁₇H₂₁NO
MW: 291.82 g/mol
Melting Pt: 168…169 °C
Storage Temperature: Ambient
MDL Number: MFCD00012479
CAS Number: 147-24-0
EINECS: 205-687-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR