Would you like to log in again?
Global Web is currently experiencing issues, technical teams are working to correct the problem.
Polyethylene Glove Bag, 69cm (27in) x 69cm (27in) x 38cm (15in), 57cm (22.5in) entrance
Supplier: Alfa Aesar
92727-LL 92727-LK
Log in to see your contract pricing and availability.
  Remember me on this device
Expand All / Collapse All
Description Dimensions Size Opening Dimensions Supplier No. VWR Catalog Number Unit Price Quantity
Glove Bag 69 x 69 x 38 cm (27 x 27 x 15") 4 bags 57 cm (22.5") 92727-LL AA92727-LL Each Retrieving Restricted
Glove Bag 69 x 69 x 38 cm (27 x 27 x 15") 1 bag 57 cm (22.5") 92727-LK AAA92727-LK Each Retrieving Restricted
restricted In order to process your orders without delay, we request that you provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:
  • State issued document with your organization's Federal Tax ID Number
  • State issued document with your organization's Resale Tax ID Number
  • City or County issued Business License
  • State Department of Health Services License
  • Any other ID issued by the State that includes the business name and address
* Please note if your account is within the State of California two of these pieces of identification will be required.
VWR will not lift restrictions for residential shipping addresses.
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Details
Select the article for which you are looking for Certificate
Indicate the Lot Number. This field is mandatory.
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty